α,2-Dichlorotoluene 98+%
|
Danger
|
Synonyms:
2-Chlorobenzyl chloride
, 1-Chloro-2-(chloromethyl)benzene
AC18653-2500
AC18653-0010
200016-500EA
29.4
USD
200016-500
200028-648
α,2-Dichlorotoluene 98+%
α,2-Dichlorotoluene
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 213 °C (1013 hPa) Melting Pt: –13 °C Density: 1.27 g/cm³ Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000893 CAS Number: 611-19-8 EINECS: 210-258-8 UN: 2235 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...