Benzimidazole 98%

Synonyms: 1H-Benzimidazole1H-Benzo[d]imidazole

AC18642-5000 AC18642-0250 AC18642-1000
200030-498EA 159 USD
200030-498 200011-334 200011-332
Benzimidazole 98%
Benzimidazole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 360 °C
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00005585
CAS Number: 51-17-2
EINECS: 200-081-4
Merck Index: 13,01082

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR