1-Methylindole 98+%

Warning

Synonyms: 1-Methyl-1H-indoleN-Methylindole

AC18630-1000 AC18630-0250 AC18630-0050
200015-850EA 31.8 USD
200015-850 103531-708 200015-848
1-Methylindole 98+%
1-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 236…239 °C (760 mmHg)
Melting Pt: 94…95 °C
Density: 1.02 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005800
CAS Number: 603-76-9
EINECS: 210-057-5
UN: 3334
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR