2,6-Dichloro-3-nitropyridine 90% tech.

Warning

AC18600-2500 AC18600-0100 AC18600-0500
200009-050EA 56.3 USD
200009-050 102777-306 200016-182
2,6-Dichloro-3-nitropyridine 90% tech.
2,6-Dichloro-3-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
MW: 192.99 g/mol
Melting Pt: 58…68 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006234
CAS Number: 16013-85-7
EINECS: 240-151-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR