1,4-Diiodobenzene 98%

Warning

AC18597-2500 AC18597-0500
103509-394EA 382 USD
103509-394 200008-504
1,4-Diiodobenzene 98%
1,4-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C (760 mmHg)
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR