2-Chloropyrimidine 98+%

Warning

AC18584-0500 AC18584-0100 AC18584-2500
200022-596EA 177 USD
200022-596 103464-560 200009-332
2-Chloropyrimidine 98+%
2-Chloropyrimidine
Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 75…76 °C (10 mmHg)
Melting Pt: 62…66 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006060
CAS Number: 1722-12-9
EINECS: 217-020-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR