4-Ethylbenzoic acid 97%

Warning

AC18449-0050 AC18449-0250
200029-806EA 24.2 USD
200029-806 200023-776
4-Ethylbenzoic acid 97%
4-Ethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002570
CAS Number: 619-64-7
EINECS: 210-605-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR