4-Bromobenzamide 97%

Warning

AC18414-0500 AC18414-0100
200050-382EA 222 USD
200050-382 200020-306
4-Bromobenzamide 97%
4-Bromobenzamide
Formula: C₇H₆BrNO
MW: 200.03 g/mol
Boiling Pt: 328 °C (1013 hPa)
Melting Pt: 189…194 °C
MDL Number: MFCD00007991
CAS Number: 698-67-9
EINECS: 211-817-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR