Cyclobutylméthanol 99%

Warning

Synonyms: CyclobutylmethanolHydroxymethylcyclobutaneCyclobutanemethanol

AC18223-0050 AC18223-0250
200024-838EA 56.6 USD
200024-838 200006-136
Cyclobutylméthanol 99%
Cyclobutylméthanol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 143…144 °C
Density: 0.91 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001330
CAS Number: 4415-82-1
EINECS: 224-575-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR