2,6-Dichlorobenzoyl chloride 99%

Danger

Synonyms: 2,6-Dichlorobenzoyl Chloride

AC18206-0100 AC18206-0500
200009-054EA 50 USD
200009-054 200004-904
2,6-Dichlorobenzoyl chloride 99%
2,6-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 142…143 °C (21 mmHg)
Melting Pt: 18 °C
Density: 1.45 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000662
CAS Number: 4659-45-4
EINECS: 225-102-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR