α,2-Dibromotoluene 98%

Danger

Synonyms: 2-Bromobenzyl bromide

AC18179-0250 AC18179-1000
200009-226EA 92.5 USD
200009-226 200009-228
α,2-Dibromotoluene 98%
α,2-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 129 °C (19 mmHg)
Melting Pt: 29…32 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000173
CAS Number: 3433-80-5
EINECS: 222-334-8
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR