α,3-Dibromotoluene 95%

Danger

Synonyms: 3-Bromobenzyl bromide1-Bromo-3-(bromomethyl)benzene

AC18178-0050 AC18178-0250
200029-210EA 112 USD
200029-210 200009-816
α,3-Dibromotoluene 95%
α,3-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 130 °C (12 mmHg)
Melting Pt: 37…42 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000176
CAS Number: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR