18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane18-Crown-6

AC18156-0250 AC18156-5000 AC18156-1000 AC18156-0050
200001-954EA 109.65 USD InStock
200001-954 103511-076 200001-956 200001-952
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%
18-Crown-6 (1,4,7,10,13,16-Hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
MW: 264.32 g/mol
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 37 - 39 °C
Density: 1.237 g/cm³ (25 °C)
Flash Pt: >110°C
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5
Merck Index: 13,02630

Order Now

Specification Test Results

Appearance Clear colorless to white Crystals or crystalline mass or liquid
Infrared spectrum Conforms
Melting point 36°C to 40°C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR