Dicyclohexylamine 99+%

Danger

AC18105-1000 AC18105-0025 AC18105-0050 AC18105-5000
200030-932EA 141 USD
200030-932 102773-852 200012-006 200006-266
Dicyclohexylamine 99+%
Dicyclohexylamine
Formula: C₁₂H₂₃N
MW: 181.32 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.91 g/cm³
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00011658
CAS Number: 101-83-7
EINECS: 202-980-7
UN: 2565
ADR: 8,III
Merck Index: 13,03122

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR