Propiononitrile 99%, Extra Pure

Danger

Synonyms: Propionitrile

AC18089-1000 AC18089-0025 AC18089-5000
103526-366EA 392 USD
103526-366 102774-482 200001-922
Propiononitrile 99%, Extra Pure
Propiononitrile
Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: –93 °C
Density: 0.77 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001948
CAS Number: 107-12-0
EINECS: 203-464-4
UN: 2404
ADR: 3,II
Merck Index: 12,08012

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR