m-Xylene 99+%, Extra Pure

Danger

Synonyms: 1,3-Dimethylbenzene

AC18086-0025 AC18086-0100 AC18086-0010
200001-916EA 146 USD
200001-916 200001-918 102777-724
m-Xylene 99+%, Extra Pure
m-Xylene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.864 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008536
CAS Number: 108-38-3
EINECS: 203-576-3
UN: 1307
ADR: 3,III
Merck Index: 12,10214

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR