2-Naphthol 99+%

Warning

Synonyms: β-Naphthol2-Hydroxynaphthaleneβ-Hydroxynaphthalene

AC18082-0100 AC18082-0025 AC18082-0010 AC18082-1000 AC18082-5000
200001-908EA 38.4 USD
200001-908 102777-214 103526-604 102776-182 200009-544
2-Naphthol 99+%
2-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 285…286 °C (760 mmHg)
Melting Pt: 120…124 °C
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00004067
CAS Number: 135-19-3
EINECS: 205-182-7
UN: 3077
ADR: 9,III
Merck Index: 12,06471

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR