1,3-Dichloro-2-propanol 99%

Danger

AC18018-2500 AC18018-0010
200040-036EA 109 USD
200040-036 200015-562
1,3-Dichloro-2-propanol 99%
1,3-Dichloro-2-propanol
Formula: ClH₂CCHOHCH₂Cl
MW: 128.99 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.351 g/cm³
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000951
CAS Number: 96-23-1
EINECS: 202-491-9
UN: 2750
ADR: 6.1,II
Merck Index: 13,03100

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR