2-Methoxythiophenol ≥97%

Warning

Synonyms: 2-Methoxybenzene-1-thiol2-Methoxybenzenethiolo-Methoxythiophenol1-Methoxy-2-mercaptobenzene2-Mercaptoanisol2-Methoxyphenyl mercaptan

AC17998-0010
103464-742EA 18.5 USD
103464-742
2-Methoxythiophenol ≥97%
2-Methoxythiophenol
Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 225…228 °C (760 mmHg)
Density: 1.14
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00004834
CAS Number: 7217-59-6
EINECS: 230-605-7
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR