5-Methoxy-1-indanone 98%

Warning

AC17967-0100 AC17967-0050 AC17967-0500
200010-742EA 181 USD
200010-742 102777-860 200017-846
5-Methoxy-1-indanone 98%
5-Methoxy-1-indanone
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 145…150 °C (0.2 mmHg)
Melting Pt: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00003789
CAS Number: 5111-70-6
EINECS: 225-838-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR