(R)-(+)-Limonene 96% unstabilized

Danger

Synonyms: D(+)-Limonene(R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene(R)-(+)-p-Mentha-1,8-dieneD-Limonene(R)-(+)-Dipentene(R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene

AC17939-0050 AC17939-5000 AC17939-0025
200004-500EA 47.19 USD
200004-500 102776-504 200001-876
(R)-(+)-Limonene 96% unstabilized
(R)-(+)-Limonene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 175.5 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.8405 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3,III
Merck Index: 13,05515

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR