1-Methylisatin 98%

Warning

Synonyms: 1-Methylindoline-2,3-dioneN-Methyl-2,3-dihydroindole-2,3-dione1-Methyl-1H-indole-2,3-dione1-Methyl-2,3-dihydroindole-2,3-dioneN-Methyl-1H-indole-2,3-dioneN-MethylisatinN-Methylindoline-2,3-dione

AC17935-0010 AC17935-0100
103511-050EA 157 USD
103511-050 200027-906
1-Methylisatin 98%
1-Methylisatin
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00005812
CAS Number: 2058-74-4
EINECS: 218-164-9
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR