Methyl-6-quinolyl ether 98%

Warning

Synonyms: 6-Methoxyquinoline

AC17934-1000 AC17934-0250
103511-048EA 183 USD
103511-048 200017-900
Methyl-6-quinolyl ether 98%
Methyl 6-quinolyl ether
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 193 °C (50 mmHg)
Melting Pt: 18…20 °C
Density: 1.152 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006800
CAS Number: 5263-87-6
EINECS: 226-077-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR