Starch, soluble for analysis

Synonyms: StarchVitex® iodine indicatorAmylodextrin

AC17713-2500 AC17713-0010
200026-816EA 104 USD
200026-816 200007-736
Starch, soluble for analysis
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Melting Pt: 256…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR