Cottonseed oil, pure

AC17711-5000 AC17711-0025
200001-850EA 31.3 USD
200001-850 103511-024
Cottonseed oil, pure
Cottonseed oil
Density: 0.91 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00130872
CAS Number: 8001-29-4
EINECS: 232-280-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR