Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy

Warning

Synonyms: Dichloromethane-[²H₂]Dideuteromethylene chlorideMethylene chloride-D2Methylene chloride-d2Dichloromethane-d2

AC17611-0050 AC17611-0250
200001-818EA 151.15 USD
200001-818 200001-820
Dichloromethane-[D₂] 99.5 atom % D for NMR spectroscopy
Dichloromethane-[D2]
Formula: CD₂Cl₂
MW: 86.92 g/mol
Boiling Pt: 40 °C (1013 hPa)
Melting Pt: –97 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR