Deuterium chloride 20% (w/w) in D₂O 100.0 atom % D for NMR spectroscopy

Warning

AC17534-0050
200057-410EA 44.43 USD
200057-410
Deuterium chloride 20% (w/w) in D₂O 100.0 atom % D for NMR spectroscopy
Deuterium chloride
Formula: DCl
MW: 37.45 g/mol
Density: 1.2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR