Silver trifluoromethanesulfonate 99+%

Warning

Synonyms: Silver trifluoromethanesulfonateTrifluoromethanesulphonic acid silver saltSilver triflateTrifluoromethanesulfonic acid silver saltSilver trifluoromethanesulphonate

AC17520-1000
103516-558EA 393 USD
103516-558
Silver trifluoromethanesulfonate 99+%
Silver trifluoromethanesulphonate
Formula: CAgF₃O₃S
MW: 256.94 g/mol
Melting Pt: 356 °C
Storage Temperature: Ambient
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR