Trifluoromethanesulfonic anhydride 98+%

Danger

Synonyms: Triflic anhydride

AC17506-0010 AC17506-2500 AC17506-0500
103529-104EA 31.4 USD
103529-104 103526-324 200001-810
Trifluoromethanesulfonic anhydride 98+%
Trifluoromethanesulphonic anhydride
Formula: C₂F₆O₅S₂
MW: 282.14 g/mol
Boiling Pt: 80 °C (760 mmHg)
Density: 1.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000408
CAS Number: 358-23-6
EINECS: 206-616-8
UN: 3265
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR