Chloroform-[D₁] 99.6+ atom % D for NMR spectroscopy

Danger

Synonyms: Chloroform-DTrichloromethane-D1Chloroform-[²H₁]

AC17488-5000
200011-636EA 383.08 USD
200011-636
Chloroform-[D₁] 99.6+ atom % D for NMR spectroscopy
Chloroform-[D1]
Formula: CDCl₃
MW: 120.38 g/mol
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR