3-Ethylaniline 98%

Danger

AC17455-2500 AC17455-0500 AC17455-0100
102777-734EA 952 USD
102777-734 200035-380 200029-284
3-Ethylaniline 98%
3-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: –8 °C
Density: 0.97 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007818
CAS Number: 587-02-0
EINECS: 209-594-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR