4-Isopropylaniline 99%

Danger

Synonyms: p-Cumidine4-Aminocumene

AC17453-0100 AC17453-0500
200014-068EA 66.5 USD
200014-068 200056-134
4-Isopropylaniline 99%
4-Isopropylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.989 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007900
CAS Number: 99-88-7
EINECS: 202-797-2
UN: 2735
ADR: 8,II
Merck Index: 13,02647

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR