4-Isopropylphenol 98%

Danger

AC17451-2500 AC17451-0050 AC17451-0010 AC17451-1000
103531-688EA 155 USD
103531-688 200023-862 200023-860 200042-072
4-Isopropylphenol 98%
4-Isopropylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 212…212.4 °C (760 mmHg)
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00002372
CAS Number: 99-89-8
EINECS: 202-798-8
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR