Phthalonitrile 98%

Danger

Synonyms: Benzene-1,2-dicarbonitrile1,2-Benzenedicarbonitrile1,2-Dicyanobenzene

AC17401-2500 AC17401-0010 AC17401-0050
200015-536EA 41.3 USD
200015-536 200033-688 103531-684
Phthalonitrile 98%
Phthalonitrile
Formula: C₈H₄N₂
MW: 128.13 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 138…142 °C
Density: 1.24 g/cm³
Flash Pt: >130 °C
Storage Temperature: Ambient
MDL Number: MFCD00001771
CAS Number: 91-15-6
EINECS: 202-044-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR