4-Nitro-m-xylene ≥99%

Synonyms: 1,3-Dimethyl-4-nitrobenzene2,4-Dimethyl-1-nitrobenzene

AC17387-1000
200036-036EA 38.1 USD
200036-036
4-Nitro-m-xylene ≥99%
4-Nitro-m-xylene
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.135 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007169
CAS Number: 89-87-2
EINECS: 201-947-4
UN: 1665
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR