4-Bromo-1-butene ≥98%

Supplier: ACROS ORGANICS MS

Danger

AC17355-0500 AC17355-0050 AC17355-0250
103526-312EA 464.8 USD InStock
103526-312 200010-234 200001-770
4-Bromo-1-butene ≥98%
4-Bromo-1-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 98°C - 100°C (760mmHg)
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 1°C
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4595 to 1.4635 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR