ε-Hexalactone monomer 99%

Warning

Synonyms: ε-CaprolactoneHexan-6-olideOxepan-2-one

AC17344-2500 AC17344-0010
200001-768EA 75 USD
200001-768 200006-434
ε-Hexalactone monomer 99%
ε-Hexalactone
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.03 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003267
CAS Number: 502-44-3
EINECS: 207-938-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR