2-Bromo-p-xylene 97%

Warning

Synonyms: 2-Bromo-1,4-dimethylbenzene2,5-Dimethylbromobenzene

AC17342-1000 AC17342-0250
200022-502EA 35.9 USD
200022-502 200016-428
2-Bromo-p-xylene 97%
2-Bromo-p-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.34 g/cm³
Flash Pt: 79 °C
MDL Number: MFCD00000074
CAS Number: 553-94-6
EINECS: 209-055-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR