Fluorescein disodium salt, pure

Synonyms: Yellow No. 82-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium saltDisodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium saltAcid Yellow 73Fluorescein sodium salt

AC17324-0025
103516-552EA 353 USD
103516-552
Fluorescein disodium salt, pure
Fluorescein disodium salt
Formula: C₂₀H₁₀Na₂O₅
MW: 376.28 g/mol
Melting Pt: 320 °C
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://us.vwr.com/stibo/web/std.lang.all/57/66/2375766.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR