2-Hydroxyisobutyric acid 98%

Danger

Synonyms: 2-Hydroxy-2-methylpropanoic acid2-Hydroxy-2-methylpropionic acidα-Hydroxyisobutyric acid

AC17310-1000 AC17310-0250 AC17310-0010
200009-428EA 33.6 USD
200009-428 103464-716 200005-072
2-Hydroxyisobutyric acid 98%
2-Hydroxyisobutyric acid
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 84 °C (1.5 mmHg)
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00004459
CAS Number: 594-61-6
EINECS: 209-848-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR