(+)-Dimethyl-L-tartrate 99%

Synonyms: (R,R)-(+)-Tartaric acid dimethyl ester(+)-Dimethyl L-tartrateL(+)-Tartaric acid dimethyl esterDimethyl (2R,3R)-2,3-dihydroxysuccinate(R,R)-(+)-Dimethyl tartrate

AC17249-1000 AC17249-0250
200015-216EA 84.9 USD
200015-216 200027-228
(+)-Dimethyl-L-tartrate 99%
(+)-Dimethyl L-tartrate
Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 157…159 °C (15hPa)
Melting Pt: 48…50 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064437
CAS Number: 608-68-4
EINECS: 210-166-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR