3-Nitrophenol 99%

Danger

Synonyms: m-Nitrophenol

AC17230-2500 AC17230-0500 AC17230-0100
200005-288EA 180 USD
200005-288 103526-304 200005-290
3-Nitrophenol 99%
3-Nitrophenol
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 194 °C (70 mmHg)
Melting Pt: 94…98 °C
Density: 1.49 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007240
CAS Number: 554-84-7
EINECS: 209-079-8
UN: 1663
ADR: 6.1,III
Merck Index: 12,06682

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR