DL-Leucine 99+%

Synonyms: 2-Amino-4-methylpentanoic acid(±)-Leucine

AC17213-0250 AC17213-2500
200012-218EA 46.1 USD
200012-218 200025-202
DL-Leucine 99+%
DL-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 293…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00063087
CAS Number: 328-39-2
EINECS: 206-328-2
Merck Index: 13,05470

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR