DL-Tryptophan 98%

Synonyms: (+/-)-2-Amino-3-(3-indolyl)propionic acid2-Amino-3-(1H-indol-3-yl)propanoic acidH-DL-Trp-OH

AC17211-1000 AC17211-0250
200037-282EA 180 USD
200037-282 200012-256
DL-Tryptophan 98%
DL-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 289…290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064339
CAS Number: 54-12-6
EINECS: 200-194-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR