DL-Valine 99+% for biochemistry

Synonyms: (±)-ValineH-DL-Val-OH2-Amino-3-methylbutanoic acid

AC17210-0500 AC17210-2500
200018-968EA 103 USD
200018-968 200012-260
DL-Valine 99+% for biochemistry
DL-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 283.5…285 °C (dec.)
Density: 1.31 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR