p-Toluenesulfonic acid monohydrate 97.5%, pure

Danger

Synonyms: 4-Methylbenzenesulphonic acid hydrate4-Methylbenzenesulfonic acid hydrateToluene-4-sulphonic acid monohydrateToluene-4-sulfonic acid monohydrate

AC17178-0050 AC17178-0025 AC17178-0010 AC17178-5000
200001-730EA 35 USD
200001-730 200014-266 200007-524 102775-906
p-Toluenesulfonic acid monohydrate 97.5%, pure
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 100…108 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8,III
Merck Index: 12,09671

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR