3-Cyclohexylpropionic acid 99%

Warning

Synonyms: 3-Cyclohexylpropanoic acid3-Cyclohexylpropionic Acid

AC17145-2500 AC17145-0500
200018-612EA 431 USD
200018-612 200024-842
3-Cyclohexylpropionic acid 99%
3-Cyclohexylpropionic acid
Formula: C₉H₁₆O₂
MW: 156.22 g/mol
Boiling Pt: 275.7 °C (1013 hPa)
Melting Pt: 14…17 °C
Density: 0.998 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001527
CAS Number: 701-97-3
EINECS: 211-861-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR