6-Chloropurine 99+%

Warning

Synonyms: 6-Chloro-9H-purine

AC17128-0050
200010-800EA 72.2 USD
200010-800
6-Chloropurine 99+%
6-Chloropurine
Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6
Merck Index: 13,02180

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR