1,2-Naphthoquinone 95% tech.

Warning

Synonyms: β-Naphthoquinone

AC17123-0250 AC17123-0010 AC17123-0050 AC17123-0100
102774-112EA 81.2 USD
102774-112 102775-494 200045-680 200045-682
1,2-Naphthoquinone 95% tech.
1,2-Naphthoquinone
Formula: C₁₀H₆O₂
MW: 158.16 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00001698
CAS Number: 524-42-5
EINECS: 208-360-2
ADR: 6.1,II
Merck Index: 12,06481

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR