1,3,5-Triisopropylbenzene 95%

AC17122-1000 AC17122-0250
103510-970EA 36 USD
103510-970 200008-442
1,3,5-Triisopropylbenzene 95%
1,3,5-Triisopropylbenzene
Formula: C₁₅H₂₄
MW: 204.36 g/mol
Boiling Pt: 232…236 °C (760 mmHg)
Density: 0.84 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00008890
CAS Number: 717-74-8
EINECS: 211-941-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR